Learn More
PDK1 Rabbit anti-Human, DyLight 488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PDK1.
Brand: Abnova PAB15335.100uL
Description
Pyruvate dehydrogenase (PDH) is a mitochondrial multienzyme complex that catalyzes the oxidative decarboxylation of pyruvate and is one of the major enzymes responsible for the regulation of homeostasis of carbohydrate fuels in mammals. The enzymatic activity is regulated by a phosphorylation/dephosphorylation cycle. Phosphorylation of PDH by a specific pyruvate dehydrogenase kinase (PDK) results in inactivation. [provided by RefSeq
Specifications
| PDK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PDK1. | |
| In 50mM sodium borate | |
| PDK1 | |
| A synthetic peptide corresponding to amino acids 300-400 of human PDK1. | |
| RUO | |
| 5163 | |
| Human | |
| Liquid |
| Western Blot | |
| DyLight 488 | |
| Western Blot (0.5-2.0 ug/ml) The optimal working dilution should be determined by the end user. | |
| PDK1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody reacts with human. No other species have been tested. The immunogen used for this antibody production has 100% homology with mouse, rat, and monkey PDK1 protein. This is specific to PDK1. By Western blot a band is seen at ∼49 KDa, representing PDK1 protein. There are also two higher and one lower non-specific molecular weight bands. | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only