Learn More
PDGFC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant PDGFC.
Brand: Abnova PAB18884.200uL
Description
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a core motif of eight cysteines. This gene product appears to form only homodimers. It differs from the platelet-derived growth factor alpha and beta polypeptides in having an unusual N-terminal domain, the CUB domain. [provided by RefSeq
Specifications
| PDGFC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant PDGFC. | |
| In buffer containing 0.02% sodium azide, 2% BSA | |
| FALLOTEIN/SCDGF | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:80000) Western Blot (1:200-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| PDGFC | |
| PDGFC | |
| Recombinant protein corresponding to 158 amino acids at C-terminus of human PDGFC. | |
| RUO | |
| 56034 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only