Learn More
PDE4D Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PDE4D.
Brand: Abnova PAB28329.100uL
Description
This gene encodes one of four mammalian counterparts to the fruit fly 'dunce' gene. The encoded protein has 3',5'-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins
Sequence: FELTLEEDGESDTEKDSGSQVEEDTSCSDSKTLCTQDSESTEIPLDEQVEEEAVGSpecifications
| PDE4D | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PDE4D. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| DPDE3/HSPDE4D/PDE4DN2/STRK1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5144 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Immunofluorescence (1-4 ug/ml) The optimal working dilution should be determined by the end user. | |
| PDE4D | |
| PDE4D | |
| Recombinant protein corresponding to amino acids of recombinant PDE4D. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only