Learn More
PDCD5 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PDCD5.
Brand: Abnova PAB16700.100ug
Description
This gene encodes a protein expressed in tumor cells during apoptosis independent of the apoptosis-inducing stimuli. Prior to apoptosis induction, this gene product is distributed in both the nucleus and cytoplasm. Once apoptosis is induced, the level of this protein increases and by relocation from the cytoplasm, it accumulates in the nucleus. Although its exact function is not defined, this protein is thought to play an early and universal role in apoptosis. [provided by RefSeq
Specifications
| PDCD5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PDCD5. | |
| In PBS (0.02% sodium azide) | |
| MGC9294/TFAR19 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| PDCD5 | |
| PDCD5 | |
| A synthetic peptide corresponding to N-terminus 13 amino acids from of human PDCD5. | |
| RUO | |
| 9141 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only