Learn More
PCTK1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PCTK1.
Brand: Abnova PAB28420.100uL
Description
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It may play a role in signal transduction cascades in terminally differentiated cells. This gene is thought to escape X inactivation. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: MDRMKKIKRQLSMTLRGGRGIDKTNGAPEQIGLDESGGGGGSDPGEAPTRAAPGELRSARGPLSSAPEIVHEDLKMGSDGESDQASATSSDEVQSPVRVRMRNHPPRKISTEDINKRLSLPADSpecifications
| PCTK1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PCTK1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| FLJ16665/PCTAIRE/PCTAIRE1/PCTGAIRE | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5127 | |
| Store at 4 °C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:100-1:250) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| PCTK1 | |
| PCTK1 | |
| Recombinant protein corresponding to human PCTK1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only