Learn More
PCSK5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PCSK5.
Brand: Abnova PAB22504.100uL
Description
The protein encoded by this gene belongs to the subtilisin-like proprotein convertase family. The members of this family are proprotein convertases that process latent precursor proteins into their biologically active products. This encoded protein mediates posttranslational endoproteolytic processing for several integrin alpha subunits. It is thought to process prorenin, pro-membrane type-1 matrix metalloproteinase and HIV-1 glycoprotein gp160. Two alternatively spliced transcripts are described for this gene but only one has its full length nature known. [provided by RefSeq
Sequence: YLEHVVVRITITHPRRGDLAIYLTSPSGTRSQLLANRLFDHSMEGFKNWEFMTIHCWGERAAGDWVLEVYDTPSQLRNFKTPGKLKEWSLVLYGTSVQPYSPTNEFPKVERFRYSRVEDPTDDSpecifications
| PCSK5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PCSK5. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PC5/PC6/PC6A/SPC6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5125 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| PCSK5 | |
| PCSK5 | |
| Recombinant protein corresponding to amino acids of human PCSK5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only