Learn More
PCDHB4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PCDHB4.
Brand: Abnova PAB20726.100uL
Description
This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3' exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. [provided by RefSeq]
Sequence: LCGPIEPCVLHFQVFLEMPVQFFQGELLIQDINDHSPIFPEREVLLKILENSQPGTLFPLLIAEDLDVGSNGLQKYTISPNSHFHILTRNHSEGKKYPDLVSpecifications
| PCDHB4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PCDHB4. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| PCDH-BETA4 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 56131 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| PCDHB4 | |
| PCDHB4 | |
| Recombinant protein corresponding to amino acids of human PCDHB4. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only