Learn More
PAX2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PAX2.
Brand: Abnova PAB18768.100ug
Description
PAX2 encodes paired box gene 2, one of many human homologues of the Drosophila melanogaster gene prd. The central feature of this transcription factor gene family is the conserved DNA-binding paired box domain. PAX2 is believed to be a target of transcriptional supression by the tumor supressor gene WT1. Mutations within PAX2 have been shown to result in optic nerve colobomas and renal hypoplasia. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Specifications
| PAX2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PAX2. | |
| In buffer containing 0.02% sodium azide | |
| PAX2 | |
| A synthetic peptide corresponding to 14 amino acids at C-terminus of human PAX2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:20000-1:80000) Western Blot (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| PAX2 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 5076 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only