Learn More
PARP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PARP1.
Brand:  Abnova PAB4469.400uL
Product Code. 16079067
  • £332.00 / 400µL
Description
This gene encodes a chromatin-associated enzyme, poly(ADP-ribosyl)transferase, which modifies various nuclear proteins by poly(ADP-ribosyl)ation. The modification is dependent on DNA and is involved in the regulation of various important cellular processes such as differentiation, proliferation, and tumor transformation and also in the regulation of the molecular events involved in the recovery of cell from DNA damage. In addition, this enzyme may be the site of mutation in Fanconi anemia, and may participate in the pathophysiology of type I diabetes. [provided by RefSeq
Specifications
Specifications
PARP1
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of PARP1.
In PBS (0.09% sodium azide)
ADPRT/ADPRT1/PARP/PARP-1/PPOL/pADPRT-1
Rabbit
Ammonium sulfate precipitation
RUO
142
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user.
PARP1
PARP1
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PARP1.
400 μL