Learn More
PARK7 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PARK7.
Brand: Abnova PAB4487.400uL
Description
The product of this gene belongs to the peptidase C56 family of proteins. It acts as a positive regulator of androgen receptor-dependent transcription. It may also function as a redox-sensitive chaperone, as a sensor for oxidative stress, and it apparently protects neurons against oxidative stress and cell death. Defects in this gene are the cause of autosomal recessive early-onset Parkinson disease 7. Two transcript variants encoding the same protein have been identified for this gene. [provided by RefSeq
Specifications
| PARK7 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PARK7. | |
| In PBS (0.09% sodium azide) | |
| DJ-1/DJ1/FLJ27376/FLJ34360/FLJ92274 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 11315 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| PARK7 | |
| PARK7 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PARK7. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only