Learn More
PANX1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PANX1.
Brand: Abnova PAB25673.100ug
Description
The protein encoded by this gene belongs to the innexin family. Innexin family members are the structural components of gap junctions. This protein and pannexin 2 are abundantly expressed in central nerve system (CNS) and are coexpressed in various neuronal populations. Studies in Xenopus oocytes suggest that this protein alone and in combination with pannexin 2 may form cell type-specific gap junctions with distinct properties. [provided by RefSeq
Specifications
| PANX1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| PANX1 | |
| PANX1 | |
| A synthetic peptide corresponding to 17 amino acids at N-terminus of human PANX1. | |
| 100 μg | |
| Primary | |
| Two transcript variants encoding different isoforms have been found for this gene. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PANX1. | |
| In PBS (0.05% sodium azide) | |
| MGC21309/MRS1/PX1/UNQ2529 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 24145 | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only