missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK1/PAK2/PAK3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PAK1/PAK2/PAK3.
Brand: Abnova PAB18202.100ug
This item is not returnable.
View return policy
Specifications
| PAK1/PAK2/PAK3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
| MGC130000/MGC130001/PAKalpha | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 5058/5062/5063 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of PAK1/PAK2/PAK3. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PAK1 | |
| A synthetic peptide corresponding to residues surrounding T423/T402/T421 of human PAK1/PAK2/PAK3. | |
| 100 μg | |
| Primary | |
| This antibody is specific to PAK1/PAK2/PAK3. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction