missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PAK1/PAK2/PAK3 (phospho T423/T402/T421), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1/PAK2/PAK3.
Brand: Abnova PAB25822.100ug
This item is not returnable.
View return policy
Description
Sequence: R-S-Tp-M-VSpecifications
| PAK1/PAK2/PAK3 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user. | |
| MGC130000/MGC130001/PAKalpha | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5058/5062/5063 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of PAK1/PAK2/PAK3. | |
| In PBS (without Mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| PAK1 | |
| Synthetic phosphopeptide corresponding to residues surrounding T423/T402/T421 of human PAK1/PAK2/PAK3. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction