Learn More
P2RY4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of P2RY4.
Brand: Abnova PAB26534.50ug
Description
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to uridine nucleotides, partially responsive to ATP, and not responsive to ADP. [provided by RefSeq
Specifications
| P2RY4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of P2RY4. | |
| In PBS (0.09% sodium azide) | |
| NRU/P2P/P2Y4/UNR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 5030 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| P2RY4 | |
| P2RY4 | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human P2RY4. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only