Learn More
P2RY13 Rabbit anti-Bovine, Human, Monkey, Pig, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of P2RY13.
Brand: Abnova PAB26458.50ug
Description
The product of this gene belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is activated by ADP. [provided by RefSeq
Specifications
| P2RY13 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of P2RY13. | |
| In PBS (0.09% sodium azide) | |
| FKSG77/GPCR1/GPR86/GPR94/P2Y13/SP174 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 53829 | |
| Bovine, Human, Monkey, Pig, Rabbit | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (8-11 ug/mL) The optimal working dilution should be determined by the end user. | |
| P2RY13 | |
| P2RY13 | |
| A synthetic peptide corresponding to 16 amino acids at 2nd extracellular domain of human P2RY13. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only