Learn More
OTX2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OTX2.
Brand: Abnova PAB19983.100uL
Description
This gene encodes a member of the bicoid sub-family of homeodomain-containing transcription factors. The encoded protein acts as a transcription factor and may play a role in brain and sensory organ development. A similar protein in mice is required for proper forebrain development. Two transcript variants encoding distinct isoforms have been identified for this gene. Other alternative splice variants may exist, but their full length sequences have not been determined. [provided by RefSeq
Sequence: SSSAPVSIWSPASISPLSDPLSTSSSCMQRSYPMTYTQASGYSQGYAGSTSYFGGMDCGSYLTPMHHQLPGPGATLSPMGTNAVTSHLNQSPASLSTQGYGASSLGFNSTTDCLDYKDQTASWKLNFNADCLDYKSpecifications
| OTX2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OTX2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MCOPS5/MGC45000 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 5015 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| OTX2 | |
| OTX2 | |
| Recombinant protein corresponding to amino acids of human OTX2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only