Learn More
OTUD6B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OTUD6B.
Brand: Abnova PAB21947.100uL
Description
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin (MIM 191339) linkages, negating the action of ubiquitin ligases. DUBA5 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.[supplied by OMIM
Sequence: IQGMKNAVPKNDKKRRKQLTEDVAKLEKEMEQKHREELEQLKLTTKENKIDSVAVNISNLVSpecifications
| OTUD6B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OTUD6B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CGI-77/DUBA5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51633 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| OTUD6B | |
| OTUD6B | |
| Recombinant protein corresponding to amino acids of human OTUD6B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only