missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OSTC.
Brand: Abnova PAB28171.50ug
This item is not returnable.
View return policy
Specifications
| OSTC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of OSTC. | |
| In PBS (0.09% sodium azide) | |
| DC2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 58505 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/ml) | |
| OSTC | |
| OSTC | |
| A synthetic peptide corresponding to 17 amino acids at N-terminus region of human OSTC. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction