Learn More
OSBPL2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OSBPL2.
Brand: Abnova PAB27896.100uL
Description
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. This encoded protein contains only the sterol-binding domain. In vitro studies have shown that the encoded protein can bind strongly to phosphatic acid and weakly to phosphatidylinositol 3-phosphate, but cannot bind to 25-hydroxycholesterol. The protein associates with the Golgi apparatus. Transcript variants encoding different isoforms have been described. [provided by RefSeq
Sequence: MNGEEEFFDAVTGFDSDNSSGEFSEANQKVTGMIDLDTSKNNRIGKTGERPSQENGIQKHRTSpecifications
| OSBPL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OSBPL2. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| FLJ20223/KIAA0772/MGC4307/MGC8342/ORP-2/ORP2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9885 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:100-1:250) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| OSBPL2 | |
| OSBPL2 | |
| Recombinant protein corresponding to amino acids of human OSBPL2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only