Learn More
OS9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OS9.
Brand: Abnova PAB12082.100uL
Description
This gene encodes a protein that is highly expressed in osteosarcomas. This protein binds to the hypoxia-inducible factor 1 (HIF-1), a key regulator of the hypoxic response and angiogenesis, and promotes the degradation of one of its subunits. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq
Specifications
| OS9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of OS9. | |
| In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.01% sodium azide) | |
| OS9 | |
| A synthetic peptide corresponding to amino acids 300-400 of human OS9. | |
| RUO | |
| 10956 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunofluorescence (0.2-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:250) Western Blot (0.2-1 ug/mL) The optimal working dilution should be determined by the end user | |
| OS9 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to isoforms 1 and 2 of human OS-9. | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only