Learn More
OR51E2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OR51E2.
Brand: Abnova PAB26597.50ug
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Specifications
| OR51E2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of OR51E2. | |
| In PBS (0.09% sodium azide) | |
| OR51E3P/OR52A2/PSGR | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 81285 | |
| Hamster, Human, Monkey, Pig, Rabbit | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (16 ug/mL) The optimal working dilution should be determined by the end user. | |
| OR51E2 | |
| OR51E2 | |
| A synthetic peptide corresponding to 14 amino acids from N-terminal extracellular domain of human OR51E2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only