Learn More
OR4K1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OR4K1.
Brand: Abnova PAB22222.100uL
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Sequence: TVDLPFCGPNEVDSFFCDLPLVIELACMDTYEMEIMTLTNSpecifications
| OR4K1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OR4K1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| OR14-19 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 79544 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| OR4K1 | |
| OR4K1 | |
| Recombinant protein corresponding to amino acids of human OR4K1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only