missing translation for 'onlineSavingsMsg'
Learn More

OR2W3, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant OR2W3.

Brand:  Abnova PAB24430.100uL

Product Code. 16044797

  • £535.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq

Sequence: GHHEVDHFLREMPALIRMACVSTVAIE
Specifications

Specifications

OR2W3
Polyclonal
Rabbit polyclonal antibody raised against recombinant OR2W3.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
OR2W3P/OR2W8P/OST718
Rabbit
Antigen affinity purification
RUO
343171
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user.
OR2W3
OR2W3
Recombinant protein corresponding to amino acids of human OR2W3.
100 μL
Primary
Human
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only