Learn More
OPTC, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OPTC.
Brand: Abnova PAB22838.100uL
Description
Opticin belongs to class III of the small leucine-rich repeat protein (SLRP) family. Members of this family are typically associated with the extracellular matrix. Opticin is present in significant quantities in the vitreous of the eye and also localizes to the cornea, iris, ciliary body, optic nerve, choroid, retina, and fetal liver. Opticin may noncovalently bind collagen fibrils and regulate fibril morphology, spacing, and organization. The opticin gene is mapped to a region of chromosome 1 that is associated with the inherited eye diseases age-related macular degeneration (AMD) and posterior column ataxia with retinosa pigmentosa (AXPC1). [provided by RefSeq
Sequence: RIDLSNNLISSIDNDAFRLLHALQDLILPENQLEALPVLPSGIEFLDVRLNRLQSSGIQPAAFRAMEKLQFLYLSDNLLDSIPSpecifications
| OPTC | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OPTC. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| OPT | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26254 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| OPTC | |
| OPTC | |
| Recombinant protein corresponding to amino acids of human OPTC. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only