Learn More
OPN3 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OPN3.
Brand: Abnova PAB26963.100uL
Description
Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. In addition to the visual opsins, mammals possess several photoreceptive non-visual opsins that are expressed in extraocular tissues. This gene, opsin 3, is strongly expressed in brain and testis and weakly expressed in liver, placenta, heart, lung, skeletal muscle, kidney, and pancreas. The gene may also be expressed in the retina. The protein has the canonical features of a photoreceptive opsin protein. [provided by RefSeq
Specifications
| OPN3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of OPN3. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| ECPN | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23596 | |
| Human, Mouse | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| OPN3 | |
| OPN3 | |
| A synthetic peptide corresponding to human OPN3. | |
| 100 μL | |
| Primary | |
| OPN3 polyclonal antibody detects endogenous levels of OPN3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only