missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Olig1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant Olig1.
Brand: Abnova PAB16939.100uL
This item is not returnable.
View return policy
Specifications
| Olig1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant Olig1. | |
| In 200mM Tris, 100mM glycine, pH 8.0 | |
| Recombinant protein corresponding to Olig1. | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:3000) Immunohistochemistry (1:1000) Immunocytochemistry (1:1000) Immunoprecipitation (1:50) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| This antibody has been directly tested for reactivity in rat and mouse. specific to the ∼27 KDa Olig1 protein in Western blots. This antibody also works well for immunohistochemistry, immunocytochemistry and immunoprecipitation. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction