Learn More
OLFM1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of OLFM1.
Brand: Abnova PAB2558.400uL
Description
This gene product shares extensive sequence similarity with the rat neuronal olfactomedin-related ER localized protein. While the exact function of the encoded protein is not known, its abundant expression in brain suggests that it may have an essential role in nerve tissue. Several alternatively spliced transcripts encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| OLFM1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of OLFM1. | |
| In PBS (0.09% sodium azide) | |
| AMY/NOE1/NOELIN1/OlfA | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 10439 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| OLFM1 | |
| OLFM1 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human OLFM1. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only