Learn More
OGT Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OGT.
Brand: Abnova PAB22483.100uL
Description
O-linked N-acetylglucosamine (O-GlcNAc) transferase (OGT) catalyzes the addition of a single N-acetylglucosamine in O-glycosidic linkage to serine or threonine residues. Since both phosphorylation and glycosylation compete for similar serine or threonine residues, the two processes may compete for sites, or they may alter the substrate specificity of nearby sites by steric or electrostatic effects. The protein contains nine tetratricopeptide repeats and a putative bipartite nuclear localization signal. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Sequence: NMFPHLKKKAVIDFKSNGHIYDNRIVLNGIDLKAFLDSLPDVKIVKMKCPDGGDNADSSNTALNMPVIPMNTIAEAVIEMINRGQIQITINGFSISNGLATTSpecifications
| OGT | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OGT. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ23071/HRNT1/MGC22921/O-GLCNAC | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8473 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| OGT | |
| OGT | |
| Recombinant protein corresponding to amino acids of human OGT. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only