Learn More
OGFR, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OGFR.
Brand: Abnova PAB20975.100uL
Description
The protein encoded by this gene is a receptor for opioid growth factor (OGF), also known as [Met(5)]-enkephalin. OGF is a negative regulator of cell proliferation and tissue organization in a variety of processes. The encoded unbound receptor for OGF has been localized to the outer nuclear envelope, where it binds OGF and is translocated into the nucleus. The coding sequence of this gene contains a polymorphic region of 60 nt tandem imperfect repeat units. Several transcripts containing between zero and eight repeat units have been reported. [provided by RefSeq
Sequence: RRKVDEGAGDSAAVASGGAQTLALAGSPAPSGHPKAGHSENGVEEDTEGRTGPKEGTPGSPSETPGPSPAGPAGDEPASpecifications
| OGFR | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OGFR. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| OGFR | |
| Recombinant protein corresponding to amino acids of human OGFR. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| OGFR | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 11054 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only