Learn More
OFD1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant OFD1.
Brand: Abnova PAB22773.100uL
Description
This gene is located on the X chromosome and encodes a centrosomal protein. A knockout mouse model has been used to study the effect of mutations in this gene. The mouse gene is also located on the X chromosome, however, unlike the human gene it is not subject to X inactivation. Mutations in this gene are associated with oral-facial-digital syndrome type I and Simpson-Golabi-Behmel syndrome type 2. Many pseudogenes have been identified; a single pseudogene is found on chromosome 5 while as many as fifteen have been found on the Y chromosome. Alternatively spliced transcripts have been described for this gene but the biological validity of these transcripts has not been determined. [provided by RefSeq
Sequence: TALENEVYCNPKQSVIDRSVNGLINGNVVPCNGEISGDFLNNPFKQENVLARMVASRITNYPTAWVEGSSPDSDLEFVANTKSpecifications
| OFD1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant OFD1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| 71-7A/CXorf5/MGC117039/MGC117040/SGBS2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8481 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| OFD1 | |
| OFD1 | |
| Recombinant protein corresponding to amino acids of human OFD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only