Learn More
ODZ1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ODZ1.
Brand: Abnova PAB28599.100uL
Description
The protein encoded by this gene belongs to the tenascin family and teneurin subfamily. It is expressed in the neurons and may function as a cellular signal transducer. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: IRTISRNQAHLNDMNIYEIASPADQELYQFTVNGTHLHTLNLITRDYVYNFTYNSEGDLGAITSSNGNSVHIRRDAGGMPLWLVVPGGQVYWLTISSNGVLKRVSAQGYNLALMTYPGNTGLLATKSNENGWTTVYEYDPEGHSpecifications
| ODZ1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ODZ1. | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide) | |
| ODZ3/TEN-M1/TNM/TNM1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10178 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| ODZ1 | |
| ODZ1 | |
| Recombinant protein corresponding to amino acids of human ODZ1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only