missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O3FAR1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of O3FAR1.
Brand: Abnova PAB25988.50ug
This item is not returnable.
View return policy
Description
GPR120 is a member of the rhodopsin family of G protein-coupled receptors (GPRs) (Fredriksson et al., 2003 [PubMed 14623098]).[supplied by OMIM
Specifications
| O3FAR1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of O3FAR1. | |
| In PBS (0.09% sodium azide) | |
| BMIQ10/GPR120/GPR129/GT01/PGR4 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 338557 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) The optimal working dilution should be determined by the end user. | |
| O3FAR1 | |
| O3FAR1 | |
| A synthetic peptide corresponding to 20 amino acids at C-terminus of human O3FAR1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except TTL (30%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction