Learn More
NUP50, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NUP50.
Brand: Abnova PAB3709.400uL
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a member of the FG-repeat containing nucleoporins that functions as a soluble cofactor in importin-alpha:beta-mediated nuclear protein import. Pseudogenes of this gene are found on chromosomes 5, 6, and 14. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| NUP50 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NUP50. | |
| In PBS (0.09% sodium azide) | |
| MGC39961/NPAP60/NPAP60L | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10762 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| NUP50 | |
| NUP50 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human NUP50. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only