Learn More
NUP210, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant NUP210.
Brand: Abnova PAB12364.100uL
Description
The nuclear pore complex is a massive structure that extends across the nuclear envelope, forming a gateway that regulates the flow of macromolecules between the nucleus and the cytoplasm. Nucleoporins are the main components of the nuclear pore complex in eukaryotic cells. The protein encoded by this gene is a membrane-spanning glycoprotein that is a major component of the nuclear pore complex. [provided by RefSeq
Specifications
| NUP210 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant NUP210. | |
| In antiserum (0.09% sodium azide) | |
| FLJ22389/GP210/KIAA0906/POM210 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| recognizes mammalian, rat & Xenopus NUP210. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| NUP210 | |
| NUP210 | |
| Recombinant protein corresponding to N-terminus human NUP210. | |
| RUO | |
| 23225 | |
| Frog, Human, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only