Learn More
NUP155 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NUP155.
Brand: Abnova PAB23161.100uL
Description
Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein encoded by this gene does not contain the typical FG repeat sequences found in most vertebrate nucleoporins. Two protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq
Sequence: TPSHGIQPPAMSTPVCALGNPATQATNMSCVTGPEIVYSGKHNGICIYFSRIMGNIWDASLVVERIFKSGNREITAIESSVPCQLLSpecifications
| NUP155 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NUP155. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| KIAA0791/N155 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9631 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| NUP155 | |
| NUP155 | |
| Recombinant protein corresponding to amino acids of human NUP155. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only