missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUMB Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Numb.
Brand: Abnova PAB12086.100uL
This item is not returnable.
View return policy
Specifications
| NUMB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Numb. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| m-numb/mnb | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to all four isoforms of the Numb protein. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (0.2-0.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| Numb | |
| Numb | |
| A synthetic peptide corresponding to C-terminus amino acids 600-653 of mouse Numb. | |
| RUO | |
| 18222 | |
| Chicken, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction