Learn More
NTRK1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NTRK1.
Brand: Abnova PAB24825.100uL
Description
This gene encodes a member of the neurotrophic tyrosine kinase receptor (NTKR) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. The presence of this kinase leads to cell differentiation and may play a role in specifying sensory neuron subtypes. Mutations in this gene have been associated with congenital insensitivity to pain, anhidrosis, self-mutilating behavior, mental retardation and cancer. Alternate transcriptional splice variants of this gene have been found, but only three have been characterized to date. [provided by RefSeq
Specifications
| NTRK1 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| NTRK1 | |
| NTRK1 | |
| A synthetic peptide corresponding to NTRK1. | |
| 100 μL | |
| Primary | |
| NTRK1 polyclonal antibody detects endogenous levels of NTRK1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of NTRK1. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| DKFZp781I14186/MTC/TRK/TRK1/TRKA/p140-TrkA | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 4914 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only