Learn More
NTF4 Rabbit anti-Human, Monkey, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant NTF4.
Brand: Abnova PAB0709.500ug
Description
This gene is a member of a family of neurotrophic factors, neurotrophins, that control survival and differentiation of mammalian neurons. The expression of this gene is ubiquitous and less influenced by environmental signals. While knock-outs of other neurotrophins including nerve growth factor, brain-derived neurotrophic factor, and neurotrophin 3 prove lethal during early postnatal development, NTF5-deficient mice only show minor cellular deficits and develop normally to adulthood. [provided by RefSeq
Specifications
| NTF4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant NTF4. | |
| Lyophilized from PBS | |
| NT-4/5/NT4/NT5/NTF5 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 4909 | |
| Human, Monkey, Mouse, Rat | |
| Lyophilized |
| ELISA, Immunohistochemistry, Inhibition Assays, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1-10 ug/mL) ELISA (1-10 ug/mL) Western Blot (1-10 ug/mL) Inhibition assay (1-10 ug/mL in vitro 2-10 ug/mL in vivo) The optimal working dilution should be determined by the end user. | |
| NTF4 | |
| NTF4 | |
| Recombinant protein corresponding to full length human NTF4. | |
| 500 μg | |
| Primary | |
| Less than 1% cross-reactivity against NGF, recombinant human BDNF and 5% to NT3 has been shown by dot blot. | |
| Store at -20°C. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only