Learn More
NSUN5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NSUN5.
Brand: Abnova PAB21311.100uL
Description
This gene encodes a protein with similarity to p120 (NOL1), a 120kDa proliferation-associated nucleolar antigen that is a member of an evolutionarily conserved protein family. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23. Alternative splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: QLPRFVRVNTLKTCSDDVVDYFKRQGFSYQGRASSLDDLRALKGKHFLLDPLMPELLVFPAQTDLHEHPLYRAGHLILSpecifications
| NSUN5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NSUN5. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ10267/MGC986/NOL1/NOL1R/NSUN5A/WBSCR20/WBSCR20A/Ynl022cL/p120 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55695 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| NSUN5 | |
| NSUN5 | |
| Recombinant protein corresponding to amino acids of human NSUN5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only