missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NSF Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Nsf.
Brand: Abnova PAB9620.100uL
This item is not returnable.
View return policy
Specifications
| NSF | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Nsf. | |
| In 10mM HEPES, 150mM NaCl, pH 7.5 (50% glycerol, 10% BSA) | |
| AI316878/AU020090/AU067812/SKD2 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 18195 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Nsf | |
| Nsf | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus rat Nsf. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction