Learn More
NRG1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NRG1.
Brand: Abnova PAB28170.50ug
Description
The protein encoded by this gene was originally identified as a 44kDa glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. This protein is a signaling protein that mediates cell-cell interactions and plays critical roles in the growth and development of multiple organ systems. It is known that an extraordinary variety of different isoforms are produced from this gene through alternative promoter usage and splicing. These isoforms are tissue-specifically expressed and differ significantly in their structure, and thereby these isoforms are classified into types I, II, III, IV, V and VI. The gene dysregulation has been linked to diseases such as cancer, schizophrenia and bipolar disorder (BPD). [provided by RefSeq
Specifications
| NRG1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NRG1. | |
| In PBS (0.09% sodium azide) | |
| ARIA/GGF/GGF2/HGL/HRG/HRG1/HRGA/NDF/SMDF | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 3084 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/ml) | |
| NRG1 | |
| NRG1 | |
| A synthetic peptide corresponding to 15 amino acids at N-terminus region of human NRG1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only