Learn More
NR5A1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR5A1.
Brand: Abnova PAB27778.50ug
Description
The protein encoded by this gene is a transcriptional activator involved in sex determination. The encoded protein binds DNA as a monomer. Defects in this gene are a cause of XY sex reversal with or without adrenal failure as well as adrenocortical insufficiency without ovarian defect. [provided by RefSeq
Specifications
| NR5A1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NR5A1. | |
| In PBS (0.09% sodium azide) | |
| AD4BP/ELP/FTZ1/FTZF1/SF-1/SF1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2516 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-18 ug/mL) The optimal working dilution should be determined by the end user. | |
| NR5A1 | |
| NR5A1 | |
| A synthetic peptide corresponding to 17 amino acid at internal region of human NR5A1. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except TRIM23 (47%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only