Learn More
NR4A3 Rabbit anti-Dog, Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NR4A3.
Brand: Abnova PAB27793.50ug
Description
This gene encodes a member of the steroid-thyroid hormone-retinoid receptor superfamily. The encoded protein may act as a transcriptional activator. The protein can efficiently bind the NGFI-B Response Element (NBRE). Three different versions of extraskeletal myxoid chondrosarcomas (EMCs) are the result of reciprocal translocations between this gene and other genes. The translocation breakpoints are associated with Nuclear Receptor Subfamily 4, Group A, Member 3 (on chromosome 9) and either Ewing Sarcome Breakpoint Region 1 (on chromosome 22), RNA Polymerase II, TATA Box-Binding Protein-Associated Factor, 68kDa (on chromosome 17), or Transcription factor 12 (on chromosome 15). Four transcript variants encoding three distinct isoforms have been identified for this gene. [provided by RefSeq
Specifications
| NR4A3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NR4A3. | |
| In PBS (0.09% sodium azide) | |
| CHN/CSMF/MINOR/NOR1/TEC | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 8013 | |
| Canine, Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-7 ug/mL) The optimal working dilution should be determined by the end user. | |
| NR4A3 | |
| NR4A3 | |
| A synthetic peptide corresponding to 20 amino acid at C-terminus of human NR4A3. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only