Learn More
NQO1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NQO1.
Brand: Abnova PAB25084.100uL
Description
This gene is a member of the NAD(P)H dehydrogenase (quinone) family and encodes a cytoplasmic 2-electron reductase. This FAD-binding protein forms homodimers and reduces quinones to hydroquinones. This protein's enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species. Mutations in this gene have been associated with tardive dyskinesia (TD), an increased risk of hematotoxicity after exposure to benzene, and susceptibility to various forms of cancer. Altered expression of this protein has been seen in many tumors and is also associated with Alzheimer's disease (AD). Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Specifications
| NQO1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| NQO1 | |
| NQO1 | |
| A synthetic peptide corresponding to NQO1. | |
| 100 μL | |
| Primary | |
| NQO1 polyclonal antibody detects endogenous levels of NQO1 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of NQO1. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| DHQU/DIA4/DTD/NMOR1/NMORI/QR1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1728 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only