missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOTUM Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOTUM.
Brand: Abnova PAB16802.100ug
This item is not returnable.
View return policy
Specifications
| NOTUM | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOTUM. | |
| In PBS (0.02% sodium azide) | |
| NOTUM | |
| A synthetic peptide corresponding to N-terminus 19 amino acids of human NOTUM. | |
| RUO | |
| 147111 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| NOTUM | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction