Learn More
NOTCH2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOTCH2.
Brand: Abnova PAB10293.200uL
Description
This gene encodes a member of the Notch family. Members of this Type 1 transmembrane protein family share structural characteristics including an extracellular domain consisting of multiple epidermal growth factor-like (EGF) repeats, and an intracellular domain consisting of multiple, different domain types. Notch family members play a role in a variety of developmental processes by controlling cell fate decisions. The Notch signaling network is an evolutionarily conserved intercellular signaling pathway which regulates interactions between physically adjacent cells. In Drosophilia, notch interaction with its cell-bound ligands (delta, serrate) establishes an intercellular signaling pathway that plays a key role in development. Homologues of the notch-ligands have also been identified in human, but precise interactions between these ligands and the human notch homologues remain to be determined. This protein is cleaved in the trans-Golgi network, and presented on the cell surface as a heterodimer. This protein functions as a receptor for membrane bound ligands, and may play a role in vascular, renal and hepatic development. [provided by RefSeq
Specifications
| NOTCH2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOTCH2. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| AGS2/hN2 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| This antiserum is directed against human NOTCH 2. Based on sequences, we expect this antibody to react as well with rat and mouse NOTCH 2. Only one amino acid difference (conservative) occurs in mouse and rat. The immunogen epitope is only exposed after gamma secretase cleavage and is not accessible in the uncleaved form. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Sandwich ELISA (1:30000-1:90000) The optimal working dilution should be determined by the end user. | |
| NOTCH2 | |
| NOTCH2 | |
| A synthetic peptide corresponding to N-terminus the cleaved N intracellular domain (NICD) of human NOTCH2. | |
| RUO | |
| 4853 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only