Learn More
NOS2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOS2.
Brand: Abnova PAB24761.100uL
Description
Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. This gene encodes a nitric oxide synthase which is expressed in liver and is inducible by a combination of lipopolysaccharide and certain cytokines. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17. [provided by RefSeq
Specifications
| NOS2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOS2. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| HEP-NOS/INOS/NOS/NOS2A | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 4843 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| NOS2 | |
| NOS2 | |
| A synthetic peptide corresponding to C-terminus of NOS2. | |
| 100 μL | |
| Primary | |
| NOS2 polyclonal antibody detects endogenous levels of NOS2 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only