Learn More
NOS1 Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOS1.
Brand: Abnova PAB17214.100ug
Description
Nitric oxide is a reactive free radical which acts as a biologic mediator in several processes, including neurotransmission and antimicrobial and antitumoral activities. Nitric oxide is synthesized from L-arginine by nitric oxide synthases. This gene encodes a nitric oxide synthase which is highly expressed in skeletal muscle. Genetic variations in this gene are associated with infantile hypertrophic pyloric stenosis type 1
Specifications
| NOS1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOS1. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| IHPS1/NOS/nNOS | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 4842 | |
| Human, Rat | |
| Lyophilized |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| NOS1 | |
| NOS1 | |
| A synthetic peptide corresponding to amino acids at C-terminus of human NOS1. | |
| 100 μg | |
| Primary | |
| The synthetic peptide of this antibody is identical to the related rat sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water to a concentration of 500μg/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only