missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOP5/NOP58 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant NOP5/NOP58.
Brand: Abnova PAB21038.100uL
This item is not returnable.
View return policy
Description
Sequence: MLVLFETSVGYAIFKVLNEKKLQEVDSLWKEFETPEKANKIVKLKHFEKFQDTAEALAAFTALMEGKINKQLKKVLKKIVKEAHEPLAVADAKLGGVIKEKLNLSCIHSPVVNELMRGIRSQMDGLSpecifications
| NOP5/NOP58 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant NOP5/NOP58. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| HSPC120 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51602 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| NOP5/NOP58 | |
| NOP5/NOP58 | |
| Recombinant protein corresponding to amino acids of human NOP5/NOP58. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu