Learn More
NOD2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of NOD2.
Brand: Abnova PAB0265.50uL
Description
This gene is a member of the Nod1/Apaf-1 family and encodes a protein with two caspase recruitment (CARD) domains and six leucine-rich repeats (LRRs). The protein is primarily expressed in the peripheral blood leukocytes. It plays a role in the immune response to intracellular bacterial lipopolysaccharides (LPS) by recognizing the muramyl dipeptide (MDP) derived from them and activating the NFKB protein. Mutations in this gene have been associated with Crohn disease and Blau syndrome. [provided by RefSeq
Sequence: LGFLVRAKGVVPGSTAPLESpecifications
| NOD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of NOD2. | |
| In serum (0.05% sodium azide) | |
| ACUG/BLAU/CARD15/CD/CLR16.3/IBD1/NLRC2/NOD2B/PSORAS1 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| NOD2 | |
| NOD2 | |
| A synthetic peptide corresponding to amino acids 582-600 of human NOD2/CARD15. | |
| RUO | |
| 64127 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only